Boekel Stopper Hole Boring Set, Brass; 9 Borers/Set
Representative image only.
Boekel Stopper Hole Boring Set, Brass; 9 Borers/Set
Cole-Parmer | Mfr: 1601AC| Item: 0629891
Easily convert solid stopper into holed stopper
₹99,900.00INR / Set of 9
₹99,900.00INR / Set of 9
Product Description
Key Features
  • Steel punch makes boring easier
  • 2 year warranty
More About this Item
Convert your solid stopper into holed-stopper with these easy-to-use borer sets. All borers come with a steel punch to make boring easier. Borer sharpener (sold separately as part number 06298-94) keeps your borer in top condition and sharpens borers up to 25mm in diameter.
Two year warranty
Specifications
  • MaterialBrass
  • DescriptionStopper Hole Boring Set, Brass; 9 Borers/Set