Bio Plas Capsette 6030 Cassettes, White, 500/pack
Representative image only.
Bio Plas Capsette 6030 Cassettes, White, 500/pack
Bio Plas | Mfr: 6030| Item: 4862000
₹11,400.00INR / Pkg of 500
₹11,400.00INR / Pkg of 500
Product Description
Key Features
  • Fit standard stainless steel and disposable plastic base molds
  • Strategically designed slots for efficient solvent flow and drainage
  • Float and remain closed throughout processing
  • Rigid construction eliminates "chattering" during sectioning
  • Acetal copolymer plastic fabrication resists heat, solvents, decalcifying solutions, and effects of microwave processing
  • Accepted by all current automatic cassette labelers
  • Capsette cassettes accept standard stainless steel lids
Conformite Europeenne
Specifications
  • ColorWhite
  • DescriptionCassettes, White, 500/pack