Benchmark Scientific D1034-MX Prefilled 2.0 mL Tubes, 0.1 mm & 4mm Silica Beads, 1.4 mm Zirconium Beads; 50/pk
Representative image only.
Benchmark Scientific D1034-MX Prefilled 2.0 mL Tubes, 0.1 mm & 4mm Silica Beads, 1.4 mm Zirconium Beads; 50/pk
Benchmark Scientific | Mfr: D1034-MX| Item: 3627187
Compatible with all common bead homogenizers
₹49,500.00INR / Pkg of 50
₹49,500.00INR / Pkg of 50
Product Description
More About this Item
Glass, zirconium, and stainless steel homogenizer beads, available in a variety of sizes, are efficient and quick at breaking down a wide range of samples in order to release the DNA, RNA, and proteins contained within cells. The beads come in prefilled unskirted tubes or in bulk, and are compatible with all popular bead homogenizers.
Glass (Silica) Beads are acid washed and processed to remove nucleases and proteases. Glass beads are ideal for homogenizing microbial applications such as algae, bacteria, fungi, mold, spores, and yeast. Zirconium Beads are quality tested to ensure the elimination of DNase, RNase, protease, and nucleic acids. The Triple-Pure™ zirconium beads are ideal for molecular biology applications.
Specifications
  • MaterialGlass and Zirconium
  • DescriptionPrefilled 2.0 mL Tubes, 0.1 mm & 4mm Silica Beads, 1.4 mm Zirconium Beads; 50/pk