Benchmark Scientific D1032-30 Prefilled 2.0 mL Tubes, Triple-Pure High Impact Zirconium Beads, 3.0 mm; 50/pk
Representative image only.
Benchmark Scientific D1032-30 Prefilled 2.0 mL Tubes, Triple-Pure High Impact Zirconium Beads, 3.0 mm; 50/pk
Benchmark Scientific | Mfr: D1032-30| Item: 3627179
Compatible with all common bead homogenizers
₹42,400.00INR / Pkg of 50
₹42,400.00INR / Pkg of 50
Product Description
More About this Item
Glass, zirconium, and stainless steel homogenizer beads, available in a variety of sizes, are efficient and quick at breaking down a wide range of samples in order to release the DNA, RNA, and proteins contained within cells. The beads come in prefilled unskirted tubes or in bulk, and are compatible with all popular bead homogenizers.
Zirconium Beads are quality tested to ensure the elimination of DNase, RNase, protease, and nucleic acids. The Triple-Pure™ zirconium beads are ideal for molecular biology applications. They come in five sizes (0.1, 0.5, 1.0. 1.5, and 3.0 mm) with the ability to homogenize everything from bacteria to large tissues. A 6-mm ceria stabilized zirconium bead is also available.
Specifications
  • MaterialZirconium
  • DescriptionPrefilled 2.0 mL Tubes, Triple-Pure High Impact Zirconium Beads, 3.0 mm; 50/pk