Benchmark Scientific D1031-T21 Empty 2.0 mL Tubes with Caps and Sealing Rings; 50/pk
Representative image only.
Benchmark Scientific D1031-T21 Empty 2.0 mL Tubes with Caps and Sealing Rings; 50/pk
Benchmark Scientific | Mfr: D1031-T21| Item: 3627184
Compatible with all common bead homogenizers
₹11,200.00INR / Pkg of 50
₹11,200.00INR / Pkg of 50
Product Description
More About this Item
Glass, zirconium, and stainless steel homogenizer beads, available in a variety of sizes, are efficient and quick at breaking down a wide range of samples in order to release the DNA, RNA, and proteins contained within cells. The beads come in prefilled unskirted tubes or in bulk, and are compatible with all popular bead homogenizers.
Specifications
  • DescriptionEmpty 2.0 mL Tubes with Caps and Sealing Rings; 50/pk