Bel-Art Plastic Cover Slips, Flexible Vinyl; 1000/Pk
Representative image only.
Bel-Art Plastic Cover Slips, Flexible Vinyl; 1000/Pk
Bel-Art | Mfr: H446900000| Item: 4851020
Protect your samples
₹24,400.00INR / Pkg of 1000
₹24,400.00INR / Pkg of 1000
Product Description
Key Features
  • Constructed of flexible vinyl
More About this Item
These plastic cover slips are unbreakable and reusable. Cover slips are square and made from flexible vinyl. Packaged as a box of 1000, divided into 10 boxes of 100 covers each, with each slip separated by a protective tissue.
Specifications
  • Thickness0.007" (0.157 mm)
  • SizeSquare (22 x 22 mm)
  • DescriptionPlastic Cover Slips, flexible vinyl, 1000/pk