Bel-Art F65000-9002 Pestle, 1.5 mL, Bulk Packed; 100/Pk

Representative image only.

Bel-Art F65000-9002 Pestle, 1.5 mL, Bulk Packed; 100/Pk

Bel-Art | Mfr: F65000-9002| Item: 4444490

Process precipitates, pellets, tissue, and cells directly in the microtube!

₹28,900.00INR / Pkg of 100

₹28,900.00INR / Pkg of 100
Product Description

Includes

two AA batteries.

Key Features

  • Homogenize samples directly in their storage container
  • Obtain thorough mixtures with the tight pestle-to-microtube tolerance
  • Protect your samples—tubes and pestles are made from virgin polypropylene
  • 3 month warranty
3 Month Warranty
Specifications
  • Capacity (mL)1.5
  • MaterialPolypropylene
  • Description Pestle, 1.5 mL, Bulk Packed; 100/Pk
  • Warranty3 month
Reviews
Q&A