Bel-Art F65000-0005 Tube and Pestle, 0.5 mL, Individually Packed; 100/Pk

Representative image only.

Bel-Art F65000-0005 Tube and Pestle, 0.5 mL, Individually Packed; 100/Pk

Bel-Art | Mfr: F65000-0005| Item: 4444476

Process precipitates, pellets, tissue, and cells directly in the microtube!

₹46,800.00INR / Pkg of 100

₹46,800.00INR / Pkg of 100
Product Description

Includes

two AA batteries.

Key Features

  • Homogenize samples directly in their storage container
  • Obtain thorough mixtures with the tight pestle-to-microtube tolerance
  • Protect your samples—tubes and pestles are made from virgin polypropylene
  • 3 month warranty
3 Month Warranty
Specifications
  • Capacity (mL)0.5
  • MaterialPolypropylene
  • Description Tube and Pestle, 0.5 mL, Individually Packed; 100/Pk
  • Warranty3 month
Reviews
Q&A