Bel-Art 18204 Single-Style Burette Clamp
Representative image only.
Bel-Art 18204 Single-Style Burette Clamp
Bel-Art | Mfr: 182040000| Item: 0670310
Soft rubber pads hold burette securely without scratching
₹10,100.00INR / Each
₹10,100.00INR / Each
Buy It Together
Current Item
Bel-Art 18204 Single-Style Burette Clamp
Bel-Art 18204 Single-Style Burette Clamp
0670310
₹10,100.00INR / Each
+ + =
Total Price 109800.00 ADD 3 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Fits standard 12.5 mm (1/2") support rod
  • Chemical resistant
  • Autoclavable
More About this Item
The non-corroding polypropylene (PP) clamps hold burettes in place without covering graduations. Clamps include 3/8" and 1/2" bushings to fit any standard support rod.
Specifications
  • Clamp MaterialPolypropylene (PP)
  • Connector MaterialPolypropylene (PP)
  • Accepts Rod Diameter (in)0.375
  • DescriptionSingle-Style Burette Clamp