Bel-Art 13691 Gas Dispersion Tubes, polyethylene, 4/bg
Representative image only.
Bel-Art 13691 Gas Dispersion Tubes, polyethylene, 4/bg
Bel-Art | Mfr: 13691-0000| Item: 0661425
₹18,800.00INR / Bag of 4
₹18,800.00INR / Bag of 4
Product Description
More About this Item
Gas Dispersion Tube linear polyethylene bell-mouthed fitting has a sealed 70-ìm porosity disc.
Specifications
  • MaterialPE
  • DescriptionGas Dispersion Tubes, polyethylene, 4/bg