BarbLock® Ultra-Secure Tubing Retainer Assembly, 3/8" for 3/8" x 5/8" Tubing
Representative image only.
BarbLock® Ultra-Secure Tubing Retainer Assembly, 3/8" for 3/8" x 5/8" Tubing
BarbLock | Mfr: BL135375 | Item: 5013443
Secure, leak-free connections for critical fluid handling applications
₹720.00INR / Each
₹720.00INR / Each
Product Description
Key Features
  • Unmatched leak prevention with 360° radial crimp and compression for secure, leak-free connections
  • Works with all flexible tubing materials and Pure-Fit® SIB® fittings
  • Certified safe materials are constructed from USP Class VI, USP <87>, and ISO 10993-5 compliant resins
  • Quick and easy setup eliminates the need for solvents or adhesives
More About this Item
BarbLock® offers ultra-secure, leak-proof connections for critical fluid transfer needs. With a unique 360° radial crimp and compression design, BarbLock® eliminates traditional bonding methods and reduces the risk of leaks. Made from high-performance materials compliant with strict safety standards, it’s ideal for applications in biopharmaceuticals research. Its easy assembly process and compatibility with various tubing types make it a convenient and reliable choice for sensitive fluid handling systems.
The BarbLock® assembly is composed of a sleeve, collet, and barbed fitting. To install, slide the sleeve and collet onto the tubing, then insert the barbed fitting. Press the sleeve and collet together until you hear a 'click' to pre-lock them. Move the assembly to the end of the barbed fitting and secure it with the assembly tool. Once locked, the assembly can only be removed using the removal tool.
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.
Specifications
  • MaterialPolypropylene
  • Compatible With3/8" x 5/8" flexible tubing
  • DescriptionUltra-Secure Tubing Retainer Assembly, 3/8" for 3/8" x 5/8" Tubing