Bacharach 0024-7059 CO calibrator kit without gas
Representative image only.
Bacharach 0024-7059 CO calibrator kit without gas
Bacharach | Mfr: H0024-7059| Item: 1010589
Calibrate your CO analyzer to ensure accuracy
₹205,000.00INR / Each
₹205,000.00INR / Each
Product Description
Includes
Variable-flow regulator, fittings, and tubing.
Key Features
  • Kit includes variable-flow regulator, fittings, and tubing
  • Compatible with Carbon monoxide analyzer 10105-26
More About this Item
Effortlessly calibrate your analyzer with this calibration kit.
Calibration cup 10105-25, calibration kit 10105-89, and calibration gas 10105-90 all requred for calibration of your analyzer.
Specifications
  • Compatible WithCarbon monoxide analyzer 10105-26
  • DescriptionCO calibrator kit without gas