Aven Tools Mighty Vue Pro LED Magnifying Lamp, 5 Diopter, UV and White LEDs, 2.25x

Representative image only.

Aven Tools Mighty Vue Pro LED Magnifying Lamp, 5 Diopter, UV and White LEDs, 2.25x

Aven Tools | Mfr: 26505-ESL-XL5-UV| Item: 4870838

Large 7.5" x 6.2" crystal-clear glass lens offer distortion-free viewing

₹96,100.00INR / Each

₹96,100.00INR / Each
Product Description

Includes

Heavy-duty mounting clamp and power adapter.

Key Features

  • 360° swivel lamp head lets you inspect from any angle
  • 36" long spring-balanced arm stays set in position
  • Covered spring arm eliminates pinching hazard
  • Features 48 white (6500K) LEDs and 30 UV (400 nm) LEDs
  • Adjustable brightness control for white LEDs (1100 nm max)
  • 5-diopter lens provides 2.75x magnification
  • ESD safe coating dissipates static energy and protects sensitive components
  • 3 year warranty

More About this Item

These Mighty Vue Pro magnifying lamps feature large glass lens for easily observing an expanded viewing area. The 36" long spring-balanced arm is easy to maneuver and stays set in position. Adjust lamp head rotates up to 360° to easily view your sample—easily move using the built-in handle.

This unique magnifying lamp features both UV LEDs and standard white LEDs for versatility. Use the UV light to see hidden details that are otherwise not visible with regular white LEDs.

3 Year WarrantyConformite EuropeenneCanadian Standards AssociationETL ListedUL Listed.
Specifications
  • Magnification2.25x
  • Diopters5D
  • Viewing Area7.5" x 6.2" (19 x 15.7 cm)
  • Lens7.5" x 6.2" (19 x 15.7 cm) clear glass
  • Mounting TypeTable clamp
  • Light Source48 high-intensity white LEDs (6500K) and 30 UV LEDs (400 nm)
  • Arm Length (in)36
  • Arm Length (cm)91.4
  • Power (VAC)100 to 240
  • Power (Hz)50/60
  • ColorBlack
  • DescriptionLED Magnifying Lamp, 5 Diopter, UV and White LEDs, 2.25x
  • Warranty3 Years
Reviews
Q&A