Aven Tools Mighty Vue Pro LED Magnifying Lamp, 3 Diopter, Color Temperature Controls, 1.75x

Representative image only.

Aven Tools Mighty Vue Pro LED Magnifying Lamp, 3 Diopter, Color Temperature Controls, 1.75x

Aven Tools | Mfr: 26505-LED-XL3| Item: 4870836

Large 7.5" x 6.2" crystal-clear glass lens offer distortion-free viewing

₹60,900.00INR / Each

₹60,900.00INR / Each
Product Description

Includes

Heavy-duty mounting clamp and power adapter.

Key Features

  • 360° swivel lamp head lets you inspect from any angle
  • 36" long spring-balanced arm stays set in position
  • Covered spring arm eliminates pinching hazard
  • Adjustable color temperature (3500K to 6500 K) and brightness control (1100 lumen max)
  • Features 60 high-intensity LEDs (30 x 3500K and 30 x 6500K)
  • 3-diopter lens provides 1.75x magnification
  • 3 year warranty

More About this Item

These Mighty Vue Pro magnifying lamps feature large glass lens for easily observing an expanded viewing area. The 36" long spring-balanced arm is easy to maneuver and stays set in position. Adjust lamp head rotates up to 360° to easily view your sample—easily move using the built-in handle.

3 Year WarrantyConformite EuropeenneCanadian Standards AssociationETL ListedUL Listed.
Specifications
  • Magnification1.75x
  • Diopters3D
  • Viewing Area7.5" x 6.2" (19 x 15.7 cm)
  • Lens7.5" x 6.2" (19 x 15.7 cm) clear glass
  • Mounting TypeTable clamp
  • Light Source60 high-intensity white LEDs (30 x 3500K and 30 x 6500K); 1100 lumens
  • Arm Length (in)36
  • Arm Length (cm)91.4
  • Power (VAC)100 to 240
  • Power (Hz)50/60
  • ColorWhite
  • DescriptionLED Magnifying Lamp, 3 Diopter, Color Temperature Controls, 1.75x
  • Warranty3 Years
Reviews
Q&A