Ashcroft T7 Temperature Switch Direct Mount 4.0" Bulb 75/205F
Representative image only.
Ashcroft T7 Temperature Switch Direct Mount 4.0" Bulb 75/205F
Ashcroft | Mfr: T7 24 TS-040 75/205F| Item: 3901276
Explosion proof and waterproof for your toughest jobs
₹198,000.00INR / Each
₹198,000.00INR / Each
Product Description
Key Features
  • Quick, accurate response via SAMA Class II vapor pressure thermal system
  • Choose the switch to meet your needs—wide variety of lengths and temperature ranges available
  • Easy to operate and install—switches meet Division 1 and 2, NEMA 7 and 9, and IP66 requirements
  • 1 year warranty
More About this Item
Single set point, fixed deadband, SPDT switches feature 316 stainless steel systems for long-lasting performance—even in harsh environments. Choose direct-mounting units with rigid probes—great for use with 3/8" nominal bore thermowells in areas with high vibration or pressure. Remote-mount switches have spring-mounted capillaries with 302 stainless steel armor and include a 1/2" NPT(M) bushing adapter.
Conformite Europeenne1 Year Warranty
Specifications
  • DescriptionTemperature Switch Direct Mount 4.0" Bulb 75/205F