Ashcroft B Series Explosion Proof Pressure Switch, 10" WC, 1/4" NPT(F); Two Set Points
Representative image only.
Ashcroft B Series Explosion Proof Pressure Switch, 10" WC, 1/4" NPT(F); Two Set Points
Ashcroft | Mfr: B724B10IW| Item: 6836048
Switches are designed with a fixed set point and deadband
₹141,000.00INR / Each
for an additional
₹37,400.00INR / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services
₹141,000.00INR / Each
Buy It Together
Current Item
Ashcroft B Series Explosion Proof Pressure Switch, 10" WC, 1/4" NPT(F); Two Set Points
Ashcroft B Series Explosion Proof Pressure Switch, 10" WC, 1/4" NPT(F); Two Set Points
6836048
₹141,000.00INR / Each
+ =
Total Price 178400.00 ADD 2 ITEMS TO CART
Product Description
Key Features
  • 1 year warranty
More About this Item
Watertight general purpose pressure switches are ideal for industrial and OEM application.
Explosion proof pressure switches are designed for hazardous process and industrial applications with
ATEX and IECEx approval for hazardous locations.
Conformite Europeenne1 Year WarrantyFM ApprovedIntrinsically SafeIP 66 RatedUL Listed.
Specifications
  • Max Pressure (in WC)10
  • CaseEpoxy-coated aluminum
  • Process Connection1/4" NPT(F)
  • Electrical Connections3/4" NPT(F) conduit
  • Enclosure RatingNEMA 7, 9, IP66
  • Media CompatibilityWater-based liquids compatible with Buna-N
  • DescriptionExplosion Proof Pressure Switch, 10" WC, 1/4" NPT(F); Two Set Points