Ashcroft 3220 Hand Jack Set, 2.5" to 12" dia Gauges

Representative image only.

Ashcroft 3220 Hand Jack Set, 2.5" to 12" dia Gauges

Ashcroft | Mfr: 3220| Item: 0735960

Easily remove or reassemble pointers

₹13,800.00INR / Each

₹13,800.00INR / Each
Product Description

Includes

Gauge-pointer remover and driver.

Key Features

  • 1 year warranty

More About this Item

The Hand Jack Set works on all 2.5'' to 12'' diameter gauges. Easily remove or reassemble pointers without damaging your gauge. Simply slide the hand jack over pointer to the movement pinion and screw down to lift pointer. Set includes a gauge-pointer remover and driver; driver features two holes to reassemble single and duplex gauges.

1 Year Warranty
Specifications
  • DescriptionHand Jack Set, 2.5" to 12" diameter gauges
  • Warranty1 Year
Reviews
Q&A