Applikon Z365001225 MicroFlask Low-Evaporation Deep-Well Sandwich Cover, 24-Well
Representative image only.
Applikon Z365001225 MicroFlask Low-Evaporation Deep-Well Sandwich Cover, 24-Well
Applikon | Mfr: Z365001225| Item: 2920845
₹52,500.00INR / Each
₹52,500.00INR / Each
Product Description
Key Features
  • 1 year warranty
More About this Item
Sandwich Covers with Layers
These sandwich covers have re-usable layers. They include a stainless steel lid for rigid support, a microfiber cotton layer, ePTFE (0.3 µm pores) layer for sterility, and a soft silicone layer to seal the system. All components can handle repeated autoclaving.
1 Year Warranty
Specifications
  • DescriptionMicroFlask Low-Evaporation Deep-Well Sandwich Cover, 24-Well