Applikon Replacement Glass Bottle for Fermenation Sampling System; 60 mL
Representative image only.
Applikon Replacement Glass Bottle for Fermenation Sampling System; 60 mL
Applikon | Mfr: Z81207BT60| Item: 0191944
₹14,600.00INR / Each
₹14,600.00INR / Each
Product Description
Key Features
  • 1 year warranty
More About this Item
Sampling system easily lets you remove a 60-mL sample from your fermentation vessel without removing the headplate. Attach system to the sample pipe via 1/4" ID tubing. Includes a 60-mL glass bottle with stainless steel fittings; optional 30-mL bottle is available, as well as additional 60-mL bottles.
1 Year Warranty
Specifications
  • DescriptionReplacement Glass Bottle for Fermenation Sampling System; 60 mL