Applikon Fermentation Septum Needle; Stainless Steel
Representative image only.
Applikon Fermentation Septum Needle; Stainless Steel
Applikon | Mfr: Z81309IN02| Item: 0191920
₹200,000.00INR / Each
₹200,000.00INR / Each
Product Description
Key Features
  • 1 year warranty
More About this Item
Septum needle is for dosing liquids or adding chemicals into your fermentation culture. Needle has a 3/16" OD stainless steel pipe for optional tubing hookup.
Autoclavable.
1 Year Warranty
Specifications
  • MaterialStainless Steel
  • DescriptionFermentation Septum Needle; Stainless Steel