Applikon Fermentation Reagent Addition Bottle, 1.0 L
Representative image only.
Applikon Fermentation Reagent Addition Bottle, 1.0 L
Applikon | Mfr: Z811301920| Item: 0191991
₹101,000.00INR / Each
₹101,000.00INR / Each
Product Description
Key Features
  • 1 year warranty
More About this Item
Reagent Addition Bottles come in two sizes. Use small bottle for acids or bases; use large bottle to hold fresh media. Transfer pipes and filter included.
Autoclavable.
Photo is representative only. Cap is now green instead of red, and the SS tubing has been replaced with silicone tubing. SS tubing and red cap configuration is still available, but components must be purchased separately.
1 Year Warranty
Specifications
  • Capacity (Liters)1
  • DescriptionFermentation Reagent Addition Bottle, 1.0 L