Applikon Fermentation/Bioreactor Porous Sparger Tip for 10-mm Size Port
Representative image only.
Applikon Fermentation/Bioreactor Porous Sparger Tip for 10-mm Size Port
Applikon | Mfr: Z811303008| Item: 0191998
₹68,900.00INR / Each
₹68,900.00INR / Each
Product Description
Key Features
  • 1 year warranty
More About this Item
Porous sparger tip for the sparger found in every system. Tip should be replaced if it breaks or clogs.
1 Year Warranty
Specifications
  • DescriptionFermentation/Bioreactor Porous Sparger Tip for 10-mm Size Port