Ansell TouchNTuff® 92-616 Lightweight Disposable Nitrile Glove, L (8, 5-9); 150/Box
Representative image only.
Ansell TouchNTuff® 92-616 Lightweight Disposable Nitrile Glove, L (8, 5-9); 150/Box
Ansell | Mfr: 105133 BOX| Item: 8161046
Puncture and chemical splash resistant
₹4,500.00INR / Box of 150
₹4,500.00INR / Box of 150
Product Description
Key Features
  • Compliant with food handling requirements
  • Packaged 150 gloves per dispenser
  • Ambidextrous
More About this Item
These lightweight nitrile gloves provide you comfort and durability. Made of 100% nitrile to prevent Type 1 allergies. These gloves offer you puncture and chemical splash protection from hydrocarbons, fats, grease, and alcohols. Ideal for food processing applications. Packaged 150 gloves per dispenser to minimize packaging.Ten packages per case.
Specifications
  • Thickness (mil)3.1
  • MaterialNitrile
  • SizeLarge
  • Length (in)9 1/2
  • Length (cm)24
  • ColorBlue
  • GripTextured fingertips
  • CuffStandard
  • ComplianceFDA 21 CFR 177-2602
  • SterileNo
  • PowderedNo
  • DescriptionLightweight Disposable Nitrile Glove, L (8, 5-9); 150/Box