Ansell TouchNTuff® 92-575 Powdered Disposable Nitrile Gloves, Size M (7, 5-8); 100/Box
Representative image only.
Ansell TouchNTuff® 92-575 Powdered Disposable Nitrile Gloves, Size M (7, 5-8); 100/Box
Ansell | Mfr: 105126 BOX| Item: 8161041
Keep hands dry and protected from chemical splashes
₹3,800.00INR / Box of 100
₹3,800.00INR / Box of 100
Product Description
Key Features
  • Textured fingertips provide secure grip
  • Complies with FDA food handling requirements
  • Ambidextrous
More About this Item
These robust nitrile gloves provide added protection for food processing and heavy-duty jobs. Powdered for easier donning and moisture absorption, the gloves are ideal for transferring liquids and solids, processing samples, handling incoming good, maintenance, and applications that have food contact.
Specifications
  • Thickness (mil)5.5
  • MaterialNitrile
  • SizeMedium
  • Length (in)9 1/2
  • Length (cm)24
  • ColorBlue
  • GripTextured fingertips
  • CuffStandard
  • ComplianceFDA 21 CFR 177-2600
  • SterileNo
  • PowderedYes
  • DescriptionPowdered Disposable Nitrile Gloves, Size M (7,5-8)