Ansell Microflex® V28 Light Duty Disposable Vinyl Glove, Size S (6.5-7); 100/Box
Representative image only.
Ansell Microflex® V28 Light Duty Disposable Vinyl Glove, Size S (6.5-7); 100/Box
Ansell | Mfr: 163295 BOX| Item: 8161048
Cost-effective protection for light duty tasks
₹4,200.00INR / Box of 100
₹4,200.00INR / Box of 100
Product Description
Key Features
  • Free from natural rubber latex
  • Produced without use of DEHP
  • Ambidextrous
More About this Item
These disposable vinyl gloves are ideal for food handling, light-duty cleaning, and general-purpose use. The gloves offer a smooth finish, giving you a more natural fit. Powder-free to reduce skin irritation. Disposable—for single use only.
Specifications
  • MaterialVinyl
  • Thickness (mil)2.8
  • SizeSmall
  • Length (in)9 1/2
  • Length (cm)24
  • ColorClear
  • CuffStandard
  • ComplianceFDA 21 CFR 177-2601
  • SterileNo
  • PowderedNo
  • DescriptionLight Duty Disposable Vinyl Glove, Size S (6.5-7); 100/Box