Ansell 224 192245 Flock-Lined, Neoprene-Over-Latex Gloves, size 9, 12 pair/pk
Representative image only.
Ansell 224 192245 Flock-Lined, Neoprene-Over-Latex Gloves, size 9, 12 pair/pk
Ansell | Mfr: 224 192245| Item: 8160150
Two-tone color indicates glove wear over time
₹11,100.00INR / Pkg of 12
₹11,100.00INR / Pkg of 12
Product Description
Key Features
  • Protects against most chemicals
  • Lining absorbs perspiration for increased comfort
More About this Item
Gloves cost less than neoprene but still provide excellent protection against most chemicals. The 100% cotton flock lining serves as an extra layer of protection and absorbs perspiration for increased comfort. Recessed diamond pattern grip.
Specifications
  • MaterialNeoprene
  • Thickness (mil)28
  • Size9
  • Length (in)13
  • Length (cm)33
  • ColorBlue and Yellow
  • LiningCotton
  • GripDiamond-embossed
  • SterileNo
  • DescriptionFlock-Lined, Neoprene-Over-Latex Gloves, size 9, 12 pair/pk