Ansell 19-026 214023 Heat-Resistant, Lined Neoprene Gloves, size 10, 26"L

Representative image only.

Ansell 19-026 214023 Heat-Resistant, Lined Neoprene Gloves, size 10, 26"L

Ansell | Mfr: 19-026 214023| Item: 8160906

Combine the flexability and chemical resistance of neoprene with the comfort and heat resistance of a soft thermal liner

₹15,900.00INR / Pair of 1

₹15,900.00INR / Pair of 1

Buy It Together

Current Item

Ansell 19-026 214023 Heat-Resistant, Lined Neoprene Gloves, size 10, 26"L
Ansell 19-026 214023 Heat-Resistant, Lined Neoprene Gloves, size 10, 26"L
8160906

₹15,900.00INR / Pair of 1
+
Scienceware 13201 Biohazard Autoclave Gloves, 232C max, 13" L
0649640

₹13,100.00INR / Pair of 1
=
Total Price 29000.00 ADD 2 ITEMS TO CART

Customers Also Bought

Product Description

More About this Item

Comfortable cotton knit lining provides extra protection against punctures. Nonslip grip.

Conformite Europeenne
Specifications
  • MaterialNeoprene
  • Size10
  • Length (in)26
  • Length (cm)66
  • ColorBlack
  • LiningThermaprene™
  • SterileNo
  • DescriptionHeat-Resistant, Lined Neoprene Gloves, size 10, 26"L
Reviews
Q&A