Amprobe TIC 410A Hot Stick (Voltage Detectors)
Representative image only.
Amprobe TIC 410A Hot Stick (Voltage Detectors)
Amprobe | Mfr: TIC 410A| Item: 2003686
Available for sale only in the United States and Canada Rugged and reliable for utility, industrial, and mine safety applications
₹75,000.00INR / Each
₹75,000.00INR / Each
Product Description
Key Features
  • desinged for checking transmission lines, power distribution equipment, down power lines, fuses, and
  • load break connectors
  • Lower voltage setting for checking voltage presence in breaker panels, breakers, power outlets and wiring
  • Non-contact AC voltage detection
  • Visual and audible voltage indication
  • Self-test verifies that tester works properly
  • Use with optional hot stick for voltage higher than 1500 VAC
  • 3 year warranty
3 Year Warranty
Specifications
  • DC Max (Volts)1500
  • AC Max (Volts)1500
  • DescriptionHot Stick (Voltage Detectors)
  • Warranty3 Years