Amprobe PK-110 Electrical Test Kit; DMM, Non-Contact Voltage Detector, and Socket Tester
Representative image only.
Amprobe PK-110 Electrical Test Kit; DMM, Non-Contact Voltage Detector, and Socket Tester
Amprobe | Mfr: PK-110| Item: 2004664
Safely test voltage level, presence, and proper wiring with this handy kit
₹19,600.00INR / Each
for an additional
₹28,000.00INR / Each
for an additional
₹29,300.00INR / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services
₹19,600.00INR / Each
Buy It Together
Current Item
Amprobe PK-110 Electrical Test Kit; DMM, Non-Contact Voltage Detector, and Socket Tester
Amprobe PK-110 Electrical Test Kit; DMM, Non-Contact Voltage Detector, and Socket Tester
2004664
₹19,600.00INR / Each
+ =
Total Price 47600.00 ADD 2 ITEMS TO CART
Product Description
Key Features
  • Convenient—contains a digital multimeter, noncontact voltage detector, and socket tester with GFCI
  • DMM safety rating of CAT II-250V and can run audible continuity, diode test, and check 1.5 and 9 V batteries
  • Noncontact voltage detector is double insulated with a safety rating of CAT IV-1000V and provides a visual voltage presence alert
  • Socket tester works to test both standard and GFCI outlets to verify proper wiring by detecting five different wiring faults
  • 1 year warranty
More About this Item
Amprobe has brought together the AM-420 digital multimeter, VP-1000 noncontact voltage detector, and ST-102B socket tester with GFCI so you can quickly and safely perform electrical voltage testing—ideal for residential applications.
The DMM measures AC/DC voltage up to 250 V, DC current, and resistance. Additional features include battery testing (of 1.5 and 9 V types), data hold, audible continuity, and diode test. Easily troubleshoot light bulbs, fuses, and detect voltage.
The LED in the tip of the pen-style noncontact voltage tester glows red in the presence 90 to 1000 volts so you can quickly verify whether outlets, switches, extension cords, circuit breakers, and fuses are live—without disconnecting the electrical system.
The socket tester with GFCI detects five common wiring faults including open ground, open neutral, open hot, hot/ground reverse, and hot/neutral reverse for both standard and GFCI outlets. Three illuminated lights provide a pattern to indicate which fault, if any, exist. The legend for the light patterns is clearly labeled on the tester.
Please refer to the datasheet or individual manuals for detailed specifications on each tool.
AM-420 Digital Multimeter, VP-1000 NC Voltage Detector, and ST-102B Socket Tester with GFCI
1 Year Warranty
Specifications
  • DC Min (Volts)0.2
  • DC Max (Volts)250
  • DC Voltage Accuracy±(1.2% rdg + 2 LSD) @ 200 mV to 200 V
  • DC Min (amps)0.2
  • DC Current Accuracy±(1.2% rdg + 5 LSD) @ 200 µA to 20 mA
  • Ac Voltage Accuracy±(1.5% rdg + 4 LSD) @ 45 Hz to 400 Hz
  • ContinuityBeeps at ≤10 Ω, off at >70 Ω
  • Diode TesterOpen-circuit voltage is around 1.5V. Normal voltage is around 0.5V to 0.8V for silicon PN junction.
  • DescriptionElectrical Test Kit; DMM, Non-Contact Voltage Detector, and Socket Tester