Aluminum Sealing Film for Thermal Sealing and General Sealing Purposes, 35 µm, Nonsterile; 500/Pk
Representative image only.
Aluminum Sealing Film for Thermal Sealing and General Sealing Purposes, 35 µm, Nonsterile; 500/Pk
Cole-Parmer| Item: 6710351
Designed for storage and PCR applications – use with temperatures ranging from -80°C to 104°C
₹134,000.00INR / Pkg of 500
₹134,000.00INR / Pkg of 500
Product Description
Key Features
  • Exceptional seal across entire plate safeguards samples during transport or storage
  • Seal on a wide range of temperatures
  • Eliminates well-to-well contamination and cross-over in PCR applications
  • Can be used with 96- and 384-well PCR plates
More About this Item
Cole-Parmer® aluminum sealing film is designed for applications ranging from PCR and real-time PCR to ELISA and cell cultures. Aluminum material is ideal for light-sensitive samples.
Please Note: PCRmax® thermal cyclers and accessories have now been rebranded Cole-Parmer®. Same great products and quality, just with a new look and name.
Specifications
  • SterileNo
  • Min Temperature (° C)-80
  • Max Temperature (° C)104
  • MaterialAluminum
  • DescriptionAluminum Sealing Film for Thermal Sealing and General Sealing Purposes, 35 µm, Nonsterile; 500/Pk