Aluminum Jar Sleeve for Milling Jar 04172-08 (2.5 gal)
Representative image only.
Aluminum Jar Sleeve for Milling Jar 04172-08 (2.5 gal)
| Mfr: AA-0320| Item: 0417218
₹285,000.00INR / Each
₹285,000.00INR / Each
Product Description
Key Features
  • 3 month warranty
More About this Item
Jar sleeves increase rigidity and traction. Order HDPE milling jars separately.
3 Month Warranty
Specifications
  • Jar Capacity (Gal)2.5
  • MaterialAnodized aluminum
  • DescriptionAluminum Jar Sleeve for Milling Jar 04172-08 (2.5 gal)
  • Warranty3 Months