Allegro 9245-01 One Man Full Faceshield Supplied Air System with 50 foot Hose
Representative image only.
Allegro 9245-01 One Man Full Faceshield Supplied Air System with 50 foot Hose
Allegro | Mfr: 9245-01| Item: 8152030
Complete and convenient NIOSH-approved systems you can take any where
₹694,000.00INR / Each
₹694,000.00INR / Each
Buy It Together
Current Item
Allegro 9245-01 One Man Full Faceshield Supplied Air System with 50 foot Hose
Allegro 9245-01 One Man Full Faceshield Supplied Air System with 50 foot Hose
8152030
₹694,000.00INR / Each
+ =
Total Price 786900.00 ADD 2 ITEMS TO CART
Product Description
More About this Item
Complete, continuous-flow low-pressure respirator systems for one or two workers are NIOSH approved. These convenient systems come in one box with one part number for easy ordering. Each system has respirator(s), breathing tube(s), air hose(s) and ambient air pump.
Full-Faceshield Systems give you the convenience of a supplied air system with the protection of an impact-resistant faceshield. High impact polycarbonate lens also features anti-fog and anti-scratch coating. Faceshield features a flip-up nylon visor with a polypropylene chin guard for added strength and durability. Cotton shroud keeps debris from falling down the back of the neck. Faceshield assembly meets ANSI Z87.1-2003 for impact resistance.
Approval: NIOSH (as a system)
Specifications
  • ComplianceNIOSH; ANSI Z87.1-2003
  • ColorBlack
  • DescriptionOne Man Full Faceshield Supplied Air System with 50 foot Hose