Allegro 3001 Alcohol-free Respirator Cleaning Wipes, 100/Box
Representative image only.
Allegro 3001 Alcohol-free Respirator Cleaning Wipes, 100/Box
Allegro | Mfr: 3001| Item: 8633201
₹3,700.00INR / Box of 100
₹3,700.00INR / Box of 100
Product Description
Key Features
  • Keep respirators clean between scheduled disinfection as mandated by OSHA
More About this Item
Alcohol-Free Respirator Cleaning Pads are for use with silicone, plastic, or other surfaces where the manufacturer recommends using a nonalcohol disinfectant. Individually wrapped, box may be used as a wall-mounted dispenser. Active ingredient: benzalkonium chloride.
Specifications
  • DescriptionAlcohol-free Respirator Cleaning Wipes, 100/Box