Allegro 2000 Reusable Respirator Storage Bag, 12" x 15"; 1/Pk

Representative image only.

Allegro 2000 Reusable Respirator Storage Bag, 12" x 15"; 1/Pk

Allegro | Mfr: 2000| Item: 8640000

₹1,600.00INR / Each

₹1,600.00INR / Each
Product Description

Key Features

  • Keep respirators contamination free

More About this Item

Reusable storage bags; durable 10-mil transparent bags with zippers. Cleaning instructions are printed right on the bag for easy reference. Measure 12"W x 15"H

Specifications
  • DescriptionReusable Respirator Storage Bag, 12" x 15"; 1/Pk
Reviews
Q&A