Alconox Tergazyme 1304 Enzyme-Active Powered Detergent; 9 x 4 lb Box/Cs
Representative image only.
Alconox Tergazyme 1304 Enzyme-Active Powered Detergent; 9 x 4 lb Box/Cs
Alconox | Mfr: 1304| Item: 1777700
Biodegradable, concentrated, and free of interfering residue
₹97,900.00INR / Case of 1
₹97,900.00INR / Case of 1
Product Description
Key Features
  • Optimize performance and environmental safety
  • Order new bulk packages for added convenience
More About this Item
These high-foaming cleaning detergents are suitable for manual cleaning applications. Ideal for use on glass, metal, plastic, rubber, and other hard materials in your critical cleaning processes. All Alconox cleaners rinse completely with no interfering residue. Alconox® and Liquinox® cleaners are biodegradable. All cleaner are safe, odorless, and FDA compliant.
Tergazyme is a concentrated, anionic detergent with protease enzyme for manual and ultrasonic cleaning. Excellent for removal of proteinaceous soils, tissue, blood, and body fluids from glassware, metals, plastic, ceramic, porcelain, rubber, and fiberglass with no interfering residues. Ideal as a cleaning agent in Reverse Osmosis and Ultra-Filtration Systems. USDA authorized. Dilute: 1:100. pH 9.5
ALCOJET, ALCONOX, LIQUI-NOX, TERG-A-ZYME-Reg TM Alconox Inc.
Specifications
  • FormatPowder
  • Dilution1:100, 9.5 pH
  • ApplicationsRemoval of proteinaceous soils, tissue, blood, and body fluids from glassware, metals, plastic, ceramic, porcelain, rubber, and fiberglass
  • DescriptionEnzyme-Active Powered Detergent; 9 x 4 lb Box/Cs