Alconox Liquinox 1201-1 Critical Cleaning Liquid Detergent; 1 gal. Bottle
Representative image only.
Alconox Liquinox 1201-1 Critical Cleaning Liquid Detergent; 1 gal. Bottle
Alconox | Mfr: 1201-1| Item: 1777840
Biodegradable, concentrated, and free of interfering residue
₹18,600.00INR / Each
₹18,600.00INR / Each
Product Description
Key Features
  • Optimize performance and environmental safety
  • Order new bulk packages for added convenience
More About this Item
These high-foaming cleaning detergents are suitable for manual cleaning applications. Ideal for use on glass, metal, plastic, rubber, and other hard materials in your critical cleaning processes. All Alconox cleaners rinse completely with no interfering residue. Alconox® and Liquinox® cleaners are biodegradable. All cleaner are safe, odorless, and FDA compliant.
Liquinox is a concentrated, anionic detergent for manual and ultrasonic cleaning. Free rinsing to give you reliable results without interfering residues. Extremely mild and completely soluble in hard and soft water. USDA authorized. Dilute 1:100. pH 8.5
ALCOJET, ALCONOX, LIQUI-NOX, TERG-A-ZYME-Reg TM Alconox Inc.
Specifications
  • FormatLiquid
  • Volume (gallons)1
  • Volume (Liters)4
  • Dilution1:100, 8.5 pH
  • ApplicationsGlass, metal, plastic, rubber, and other hard materials
  • DescriptionCritical Cleaning Liquid Detergent; 1 gal. Bottle