Alconox Citranox 1801-1 Liquid Acid Cleaner and Detergent; 1 gal. Bottle
Representative image only.
Alconox Citranox 1801-1 Liquid Acid Cleaner and Detergent; 1 gal. Bottle
Alconox | Mfr: 1801-1| Item: 1777515
Biodegradable, concentrated, and free of interfering residue
₹28,400.00INR / Each
₹28,400.00INR / Each
Product Description
Key Features
  • Optimize performance and environmental safety
  • Order new bulk packages for added convenience
More About this Item
These high-foaming cleaning detergents are suitable for manual cleaning applications. Ideal for use on glass, metal, plastic, rubber, and other hard materials in your critical cleaning processes. All Alconox cleaners rinse completely with no interfering residue. Alconox® and Liquinox® cleaners are biodegradable. All cleaner are safe, odorless, and FDA compliant.
Citranox is a phosphate free, concentrated cleaner and metal brightener for high performance removal of metal oxides, scale, salts, and inorganic residues in manual and ultrasonic cleaning. Corrosion inhibited and free rinsing for reliable results and no interfering residues. Ideal for trace metal analysis. USDA authorized. Dilute 2:100. pH 2.5
ALCOJET, ALCONOX, LIQUI-NOX, TERG-A-ZYME-Reg TM Alconox Inc.
Specifications
  • FormatLiquid
  • Volume (gallons)1
  • Volume (Liters)4
  • Dilution2:100, 2.5 pH
  • ApplicationsRemoval of metal oxides, scale, salts, and inorganic residues in manual and ultrasonic cleaning
  • DescriptionLiquid Acid Cleaner and Detergent; 1 gal. Bottle