Alconox 1125 Powdered Precision Cleaner; 25 lb Box
Representative image only.
Alconox 1125 Powdered Precision Cleaner; 25 lb Box
Alconox | Mfr: 1125| Item: 1777550
Biodegradable, concentrated, and free of interfering residue
₹64,300.00INR / Carton of 1
₹64,300.00INR / Carton of 1
Product Description
Key Features
  • Optimize performance and environmental safety
  • Order new bulk packages for added convenience
More About this Item
These high-foaming cleaning detergents are suitable for manual cleaning applications. Ideal for use on glass, metal, plastic, rubber, and other hard materials in your critical cleaning processes. All Alconox cleaners rinse completely with no interfering residue. Alconox® and Liquinox® cleaners are biodegradable. All cleaner are safe, odorless, and FDA compliant.
Alconox powdered precision cleaner is a concentrated, anionic detergent for manual and ultrasonic cleaning. Free rinsing to give you reliable results without interfering residues. Ideal for cleaning contaminants from glassware, metals, plastics, ceramic, porcelain, rubber, and fiberglass. Excellent replacement for corrosive acids and hazardous solvents. USDA authorized. Dilute 1:100. pH 9.5
ALCOJET, ALCONOX, LIQUI-NOX, TERG-A-ZYME-Reg TM Alconox Inc.
Specifications
  • FormatPowder
  • Dilution1:100, 9.5 pH
  • ApplicationsGlassware, metals, plastics, ceramic, porcelain, rubber, and fiberglass
  • DescriptionPowdered Precision Cleaner; 25 lb Box