Air Impurities Removal Systems Prefilters for HEPA Filter for Air Cleaning Systems, Pack of Six
Representative image only.
Air Impurities Removal Systems Prefilters for HEPA Filter for Air Cleaning Systems, Pack of Six
Air Impurities Removal Systems | Mfr: RF_981-6| Item: 3370312
₹22,800.00INR / Pkg of 6
₹22,800.00INR / Pkg of 6
Product Description
More About this Item
High-Efficiency Filters capture smoke and dust — ideal if you mix dry chemicals, sand, grind, or do light welding or soldering. Prefilter is 30 to 35% efficient; high-efficiency cartridge is 96.5% efficient.
HEPA Filters capture soldering fumes and vapors that contain lead. Prefilter is 30 to 35% efficient; HEPA filter is 99.97% efficient.
Final Adsorption Carbon Module captures volatile solvents, touch-up paint fumes, and other noxious odors. Available in combination with primary high-efficiency or HEPA filter (33703-02 and -04), or use asa final stage for portable air cleaning system.
Specifications
  • DescriptionPrefilters for HEPA Filter for Air Cleaning Systems, Pack of Six