Air Impurities Removal Systems Prefilters for Air Cleaning Systems, 4/pk
Representative image only.
Air Impurities Removal Systems Prefilters for Air Cleaning Systems, 4/pk
Air Impurities Removal Systems | Mfr: RF-987-1| Item: 3370346
₹36,600.00INR / Pkg of 4
₹36,600.00INR / Pkg of 4
Product Description
More About this Item
Easily install or change filters in just seconds. Filter packages designed to suit your specific working conditions
High-Efficiency Filters captures smoke and dust — ideal if you mix dry chemicals, sand, grind, or do light welding. If you use solvents or paint in the same process, choose a combination high-efficiency filter with refillable 2" thick carbon module. Filter is 95% efficient.
HEPA Filters capture soldering fumes and vapors that contain lead. If you use solvents or paint in the same process, choose a combination HEPA filter with refillable 2" thick carbon module. Filter is 99.97% efficient.
Activated Carbon Filter captures volatile solvents, touch-up paint fumes, and other noxious odors. The primary
Specifications
  • DescriptionPrefilters for Air Cleaning Systems, 4/pk