Air Impurities Removal Systems HEPA Filter for Air Cleaning Systems
Representative image only.
Air Impurities Removal Systems HEPA Filter for Air Cleaning Systems
Air Impurities Removal Systems | Mfr: RF-981-5| Item: 3370309
₹96,700.00INR / Each
₹96,700.00INR / Each
Product Description
More About this Item
High-Efficiency Filters capture smoke and dust — ideal if you mix dry chemicals, sand, grind, or do light welding or soldering. Prefilter is 30 to 35% efficient; high-efficiency cartridge is 96.5% efficient.
HEPA Filters capture soldering fumes and vapors that contain lead. Prefilter is 30 to 35% efficient; HEPA filter is 99.97% efficient.
Final Adsorption Carbon Module captures volatile solvents, touch-up paint fumes, and other noxious odors. Available in combination with primary high-efficiency or HEPA filter (33703-02 and -04), or use asa final stage for portable air cleaning system.
Specifications
  • DescriptionHEPA Filter for Air Cleaning Systems