Air Impurities Removal Systems Activated Carbon Refil for 33703-42, -44
Representative image only.
Air Impurities Removal Systems Activated Carbon Refil for 33703-42, -44
Air Impurities Removal Systems | Mfr: RF-987-4C| Item: 3370348
₹31,300.00INR / Each
₹31,300.00INR / Each
Product Description
More About this Item
Easily install or change filters in just seconds. Filter packages designed to suit your specific working conditions
High-Efficiency Filters captures smoke and dust — ideal if you mix dry chemicals, sand, grind, or do light welding. If you use solvents or paint in the same process, choose a combination high-efficiency filter with refillable 2" thick carbon module. Filter is 95% efficient.
HEPA Filters capture soldering fumes and vapors that contain lead. If you use solvents or paint in the same process, choose a combination HEPA filter with refillable 2" thick carbon module. Filter is 99.97% efficient.
Activated Carbon Filter captures volatile solvents, touch-up paint fumes, and other noxious odors. The primary
Specifications
  • DescriptionActivated Carbon Refil for 33703-42, -44