AEMC 275HVD Hi-Voltage Detector Noncontact 240v/275kv
Representative image only.
AEMC 275HVD Hi-Voltage Detector Noncontact 240v/275kv
AEMC | Mfr: 2131.12| Item: 2001983
₹106,000.00INR / Each
₹106,000.00INR / Each
Product Description
Key Features
  • 1 year warranty
More About this Item
Non-contact with high voltage detector
FEATURES
  • Non-contact with high voltage
  • Wide range of detection; finds 80V to 275kV; one instrument fits all applications
  • Eight switch selectable voltage ranges (AE213112)
  • Ten switch-selectable voltage sensitivity ranges (AE213113)
  • Lightweight, minimizes sway at the end of long hot sticks
  • Self-test position ensures that all system functions and indicators are working properly by energizing the complete system
  • Audible and visual indication of live voltage
  • Automatic self-test beeps every two seconds (AE213113)
  • Digital filter rejects signals outside of 40 to 70 Hz
  • Designed for use with hot sicks incorporating a universal spline adapter
1 Year Warranty
Specifications
  • DC Max (Volts)240
  • AC Max (Volts)240
  • DescriptionHi-Voltage Detector Noncontact 240v/275kv