Advantec UNIV-R Universal pH Test Paper; pH Range 1 to 11; 9 mm x 6 m Roll
Representative image only.
Advantec UNIV-R Universal pH Test Paper; pH Range 1 to 11; 9 mm x 6 m Roll
Advantec | Mfr: UNIV-R| Item: 5920106
Determine pH easily and inexpensively without instrumentation
₹7,200.00INR / Pkg of 1
₹7,200.00INR / Pkg of 1
Buy It Together
Current Item
Advantec UNIV-R Universal pH Test Paper; pH Range 1 to 11; 9 mm x 6 m Roll
Advantec UNIV-R Universal pH Test Paper; pH Range 1 to 11; 9 mm x 6 m Roll
5920106
₹7,200.00INR / Pkg of 1
+ =
Total Price 19600.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Color changes instantaneously, simply compare to chart for determination
  • Available in two convenient formats — a booklet of 20 strips, 7 x 70 mm; or a single 9-mm wide roll in a convenient dispenser
  • Packaged as a roll measuring 9 mm x 6 m in a convenient dispenser
  • 1 year warranty
1 Year Warranty
Specifications
  • TestpH
  • Measurement Points1 to 11 pH
  • DescriptionUniversal pH Test Paper; pH Range 1 to 11; 9 mm x 6 m Roll