Advantec Three-piece monitor, plain surface, 0.80 um, 50/pack
Representative image only.
Advantec Three-piece monitor, plain surface, 0.80 um, 50/pack
Advantec | Mfr: 37AS380AN| Item: 0293506
Ideal for bacterial, yeast, mold and other contamination analysis Use 3-piece monitor for aerosol analysis
₹56,600.00INR / Pkg of 50
₹56,600.00INR / Pkg of 50
Product Description
Key Features
  • 1 year warranty
More About this Item
Pre-assembled 3-piece units contain a built-in membrane filter and a support pad. Monitors are suitable for field collection and transportation of samples as well as analysis in the laboratory. Samples collected on the press-fit membrane can then be cultured in place or removed for further analysis. Individually packaged sets are pre-sterilized.
1 Year Warranty
Specifications
  • ColorWhite
  • Port Membrane Diameter (µm)0.8
  • Surface MembranePlain
  • DescriptionThree-piece monitor, plain surface, 0.80 um, 50/pack