Advantec S045A330R Nitrocellulose membranes 0.45um 300cm/roll

Representative image only.

Advantec S045A330R Nitrocellulose membranes 0.45um 300cm/roll

Advantec | Mfr: S045A330R| Item: 0664580

Free of acetate and other esters

Call For Price

Call For Price

Buy It Together

Current Item

Advantec S045A330R Nitrocellulose membranes 0.45um 300cm/roll
Advantec S045A330R Nitrocellulose membranes 0.45um 300cm/roll
0664580

Call For Price
+ =
Total Price 144000.00 ADD 2 ITEMS TO CART
Product Description

Key Features

  • Maximum protein/nucleic acid binding
  • Low background
  • 1 year warranty

More About this Item

Available in two pore sizes—0.45 µm is suitable for most blotting applications, 0.20 µm sizes for lower molecular weights.

1 Year Warranty
Specifications
  • MaterialNitrocellulose
  • Pore Size (micron)0.45
  • Sheet Length (in)118
  • Sheet Width (in)130
  • DescriptionNitrocellulose membranes 0.45um 300cm/roll
  • Warranty1 year
Reviews
Q&A