Advantec QR10047MM Air Sampling Filters, Quartz, 47-mm dia disc; pack of 100.
Representative image only.
Advantec QR10047MM Air Sampling Filters, Quartz, 47-mm dia disc; pack of 100.
Advantec | Mfr: QR10047MM| Item: 0664952
Quartz filters ideal for sampling airborne particulates
₹35,200.00INR / Pkg of 100
₹35,200.00INR / Pkg of 100
Product Description
Key Features
  • 1 year warranty
More About this Item
Quartz Filters are used for air sampling of acidic gases at high temperatures. Quartz fibers are unaffected by humidity, highly resistant to chemical attack, and biologically inert. Filters are prefired at 1832°F (1000°C) for two hours to reduce organic contamination. Show very low trace metals content and do not absorb nitrous and sulfur dioxides.
1 Year Warranty
Specifications
  • MaterialQuartz Microfiber
  • Diameter (mm)47
  • Diameter (cm)4.7
  • DescriptionAir Sampling Filters, Quartz, 47-mm dia disc; pack of 100.