Advantec Litmus Red Test Paper, 10 Booklets of 20 Strips
Representative image only.
Advantec Litmus Red Test Paper, 10 Booklets of 20 Strips
Advantec | Mfr: 07020020| Item: 5920102
Determine pH easily and inexpensively without instrumentation
₹2,000.00INR / Pkg of 10
₹2,000.00INR / Pkg of 10
Buy It Together
Current Item
Advantec Litmus Red Test Paper, 10 Booklets of 20 Strips
Advantec Litmus Red Test Paper, 10 Booklets of 20 Strips
5920102
₹2,000.00INR / Pkg of 10
+ =
Total Price 14400.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Color changes instantaneously, simply compare to chart for determination
  • Available in two convenient formats — a booklet of 20 strips, 7 x 70 mm; or a single 9-mm wide roll in a convenient dispenser
  • Packaged as 10 booklets of 20 strips each; strips measure 7 x 70 mm
  • 1 year warranty
1 Year Warranty
Specifications
  • TestpH
  • Measurement PointsPresence/absence
  • Reagent PlatformStrips
  • Tests Per Kit200
  • DescriptionLitmus Red Test Paper, 10 Booklets of 20 Strips