Advantec GC9090MM Grade GC90 Glass Fiber Filters; 90mm dia; 100/box

Representative image only.

Advantec GC9090MM Grade GC90 Glass Fiber Filters; 90mm dia; 100/box

Advantec | Mfr: GC9090MM| Item: 0665612

For analytical or gravimetric studies

₹30,600.00INR / Pkg of 100

₹30,600.00INR / Pkg of 100
Product Description

Key Features

  • 1 year warranty

More About this Item

High wet strength.

1 Year Warranty
Specifications
  • MaterialGlass Microfiber
  • Diameter (mm)90
  • Diameter (cm)9
  • DescriptionGrade GC90 Glass Fiber Filters; 90mm dia; 100/box
  • Warranty1 year
Reviews
Q&A