Advantec GC9047MM Grade GC90 Glass Fiber Filters; 47mm dia; 100/box

Representative image only.

Advantec GC9047MM Grade GC90 Glass Fiber Filters; 47mm dia; 100/box

Advantec | Mfr: GC9047MM| Item: 0665782

For analytical or gravimetric studies

₹12,400.00INR / Pkg of 100

₹12,400.00INR / Pkg of 100
Product Description

Key Features

  • 1 year warranty

More About this Item

High wet strength.

1 Year Warranty
Specifications
  • MaterialGlass Microfiber
  • Diameter (mm)47
  • Diameter (cm)4.7
  • DescriptionGrade GC90 Glass Fiber Filters; 47mm dia; 100/box
  • Warranty1 year
Reviews
Q&A